missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine 98.0+%, TCI America™

Catalog No. B30725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30725G 5 g
B307225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30725G Supplier TCI America Supplier No. B30725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD000371
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
Synonym fmoc-lys boc-oh, fmoc-lys boc, na-fmoc-ne-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n-fmoc-n'-boc-l-lysine, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n, a-fmoc-n, a-boc-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 2724628
IUPAC Name (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 468.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine
TSCA No
Recommended Storage Refrigerator