missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98.0+%, TCI America™
$72.70 - $219.95
Chemical Identifiers
| CAS | 92122-45-7 |
|---|---|
| Molecular Formula | C26H32N2O6 |
| Molecular Weight (g/mol) | 468.55 |
| MDL Number | MFCD00065660 |
| InChI Key | UMRUUWFGLGNQLI-ANBDAQEENA-N |
| Synonym | fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 |
| PubChem CID | 13585941 |
| IUPAC Name | (2R)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 92122-45-7 | |
| 468.55 | |
| UMRUUWFGLGNQLI-ANBDAQEENA-N | |
| 13585941 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C26H32N2O6 | |
| MFCD00065660 | |
| fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 | |
| (2R)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
Specifications
| 92122-45-7 | |
| White-Yellow | |
| MFCD00065660 | |
| fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 468.55 | |
| 468.55 | |
| Crystalline Powder |
| 130°C | |
| C26H32N2O6 | |
| 1 g | |
| UMRUUWFGLGNQLI-ANBDAQEENA-N | |
| (2R)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid | |
| 13585941 | |
| ≥98.0% (HPLC,T) | |
| Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator