missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%

CAS: 2389-45-9 | C19H28N2O6 | 380.44 g/mol

$87.92 - $254.18

Chemical Identifiers

CAS 2389-45-9
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00065584
InChI Key BDHUTRNYBGWPBL-GGYSOQFKNA-N
Synonym boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
PubChem CID 2724765
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5953704
View Documents
Thermo Scientific Chemicals
H5953704
2 g
Each for $87.92
Only null left
 
AAH5953709
View Documents
Thermo Scientific Chemicals
H5953709
10 g
Each for $254.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2389-45-9
380.44
BDHUTRNYBGWPBL-GGYSOQFKNA-N
2724765
C19H28N2O6
MFCD00065584
boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

2389-45-9
MFCD00065584
1917222
BDHUTRNYBGWPBL-GGYSOQFKNA-N
(2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
2724765
95%
C19H28N2O6
2 g
boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
380.44
380.44
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-221-0

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only