missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neotame Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Neotame is a water-soluble, non-calorific, high-intensity sweetener utilized in food and beverages. Its structure resembles that of aspartame. Its sweetness potency is about 700-13000 times greater than that of sucrose and 30-60 times greater than that of aspartame. It is known as one of the sweetest artificial sweeteners.
Specifications
Specifications
| CAS | 243116-62-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C20H30N2O5 · H2O |
| Synonym | Neotame monohydrate; N-[N-(3,3-Dimethylbutyl)-L-alpha-aspartyl]-L-phenylalanine 1-methyl ester monohydrate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 396.48 |
| Grade | Certified Reference Material |