missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neostigmine Methyl Sulfate 98.0+%, TCI America™
$210.63 - $764.27
Chemical Identifiers
| CAS | 51-60-5 |
|---|---|
| Molecular Formula | C13H22N2O6S |
| Molecular Weight (g/mol) | 334.387 |
| MDL Number | MFCD00011796 |
| InChI Key | OSZNNLWOYWAHSS-UHFFFAOYSA-M |
| Synonym | (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulfate, (3-Hydroxyphenyl)trimethylammonium Methyl Sulfate Dimethylcarbamate |
| PubChem CID | 5824 |
| ChEBI | CHEBI:7516 |
| IUPAC Name | [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate |
| SMILES | CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C.COS(=O)(=O)[O-] |
Chemical Identifiers
| 51-60-5 | |
| 334.387 | |
| OSZNNLWOYWAHSS-UHFFFAOYSA-M | |
| 5824 | |
| [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate |
| C13H22N2O6S | |
| MFCD00011796 | |
| (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulfate, (3-Hydroxyphenyl)trimethylammonium Methyl Sulfate Dimethylcarbamate | |
| CHEBI:7516 | |
| CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C.COS(=O)(=O)[O-] |
Specifications
| 51-60-5 | |
| White | |
| C13H22N2O6S | |
| 5 g | |
| (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulfate, (3-Hydroxyphenyl)trimethylammonium Methyl Sulfate Dimethylcarbamate | |
| CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C.COS(=O)(=O)[O-] | |
| 334.387 | |
| CHEBI:7516 | |
| ≥98.0% (N) | |
| Neostigmine Methyl Sulfate |
| 146°C | |
| 3.0 - 5.0 | |
| MFCD00011796 | |
| 2811 | |
| OSZNNLWOYWAHSS-UHFFFAOYSA-M | |
| [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate | |
| 5824 | |
| 334.39 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0708
RTECSNumber : CY1225000
TSCA : No