missing translation for 'onlineSavingsMsg'
Learn More

Neostigmine bromide

Catalog No. AAJ62300MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62300MD 250 mg
AAJ6230003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62300MD Supplier Thermo Scientific Chemicals Supplier No. J62300MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A reversible acetylcholinesterase inhibitor | CAS: 114-80-7 | C12H19BrN2O2 | 303.20 g/mol

A reversible acetylcholinesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114-80-7
Molecular Formula C12H19BrN2O2
Molecular Weight (g/mol) 303.20
MDL Number MFCD00011795
InChI Key LULNWZDBKTWDGK-UHFFFAOYSA-M
Synonym neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide
PubChem CID 8246
ChEBI CHEBI:179557
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 250 mg
UN Number UN2811
Merck Index 14,6464
Formula Weight 303.2
Physical Form Powder
Chemical Name or Material Neostigmine bromide
Hazard Category H301-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H300-H310-H330-H334-H315-H319-H317-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-054-8
RTECSNumber BR3150000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only