missing translation for 'onlineSavingsMsg'
Learn More

Neostigmine Bromide 99.0+%, TCI America™

Catalog No. p-7133770
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03585G 5 g
N035825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03585G Supplier TCI America Supplier No. N03585G
Only null left

Chemical Identifiers

CAS 114-80-7
Molecular Formula C12H19BrN2O2
Molecular Weight (g/mol) 303.20
MDL Number MFCD00011795
InChI Key LULNWZDBKTWDGK-UHFFFAOYSA-M
Synonym neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide
PubChem CID 8246
ChEBI CHEBI:179557
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C

Specifications

Color White
Quantity 5 g
UN Number 2811
Formula Weight 303.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Neostigmine Bromide
EINECSNumber (9)-0708&(9)-1092
RTECSNumber BR3150000
TSCA No