missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neosolaniol reference material, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 36519-25-2 |
| Physical Form | Neat |
| Linear Formula | C19H26O8 |
| MDL Number | MFCD16661260 |
| Synonym | Neosolaniol from Fusarium sp.; 4 beta,15-Diacetoxy-3 alpha,8 alpha-dihydroxy-12,13-epoxytrichothec-9-ene; 8-Hydroxydiacetoxyscirpenol; NEO |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 382.40 |
| Grade | Analytical Standard |