missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neosaxitoxin-15N7 Solution, 10 μg/g, Certified Reference Material, MilliporeSigma™ Supelco™
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
Specifications
Specifications
| Product Line | TraceCERT |
| Concentration | 10 μg/g in hydrochloric acid (nom inal concentration) |
| Linear Formula | C10H1715N7O5 |
| Synonym | neo-Saxitoxin-15N7 solution; NeoSTX-15N7 |
| Quantity | 0.5 mL |
| Formula Weight | 322.24 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC