missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl Glycol Diglycidyl Ether 40.0+%, TCI America™

Catalog No. N044825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N044825ML 25 mL
N0448500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N044825ML Supplier TCI America Supplier No. N044825ML

Chemical Identifiers

CAS 17557-23-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00040687
InChI Key KUAUJXBLDYVELT-UHFFFAOYSA-N
Synonym Diglycidyloxyneopentane
PubChem CID 28594
IUPAC Name 2-[[2,2-dimethyl-3-(oxiran-2-ylmethoxy)propoxy]methyl]oxirane
SMILES CC(C)(COCC1CO1)COCC2CO2

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 216.28
Percent Purity ≥40.0% (GC)
Physical Form Liquid
Chemical Name or Material Neopentyl Glycol Diglycidyl Ether
EINECSNumber (2)-0396
RTECSNumber TX3760000
TSCA Yes