missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neopentyl Glycol Diacrylate (stabilized with MEHQ) 89.0+%, TCI America™
Supplier: TCI America N0790100G
Specifications
| Neopentyl Glycol Diacrylate (stabilized with MEHQ) | |
| 6°C | |
| 96°C | |
| MFCD00048148 | |
| 2810 | |
| MXFQRSUWYYSPOC-UHFFFAOYSA-N | |
| (2,2-dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate | |
| 16680 | |
| ≥89.0% (GC) |
| 2223-82-7 | |
| Yellow | |
| C11H16O4 | |
| 100 g | |
| 2,2-Dimethyl-1,3-propanediol Diacrylate, 1,3-Bis(acryloyloxy)-2,2-dimethylpropane | |
| CC(C)(COC(=O)C=C)COC(=O)C=C | |
| 212.245 | |
| 212.25 | |
| Liquid |
Chemical Identifiers
| 2223-82-7 | |
| 212.245 | |
| MXFQRSUWYYSPOC-UHFFFAOYSA-N | |
| 16680 | |
| CC(C)(COC(=O)C=C)COC(=O)C=C |
| C11H16O4 | |
| MFCD00048148 | |
| 2,2-Dimethyl-1,3-propanediol Diacrylate, 1,3-Bis(acryloyloxy)-2,2-dimethylpropane | |
| (2,2-dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate |
Safety and Handling
EINECSNumber : (2)-1007&(2)-0958
RTECSNumber : AS8925000
TSCA : Yes