accessibility menu, dialog, popup

UserName

CH3CH3OOCH2OOH2C

CAS RN 2223-82-7

IUPAC Name: 2,2-dimethyl-3-(prop-2-enoyloxy)propyl prop-2-enoate
Synonyms: neopentylglycol diacrylate
Molecular Weight (g/mol): 212.25
Molecular Formula: C11H16O4
InChi Key: MXFQRSUWYYSPOC-UHFFFAOYSA-N
SMILES: CC(C)(COC(=O)C=C)COC(=O)C=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results