missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl Glycol Diacrylate (stabilized with MEHQ) 89.0+%, TCI America™

Catalog No. N079025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N079025G 25 g
N0790100G 100 g
N0790500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N079025G Supplier TCI America Supplier No. N079025G
Only null left

Chemical Identifiers

CAS 2223-82-7
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.245
MDL Number MFCD00048148
InChI Key MXFQRSUWYYSPOC-UHFFFAOYSA-N
Synonym 2,2-Dimethyl-1,3-propanediol Diacrylate, 1,3-Bis(acryloyloxy)-2,2-dimethylpropane
PubChem CID 16680
IUPAC Name (2,2-dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate
SMILES CC(C)(COC(=O)C=C)COC(=O)C=C

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 2810
Formula Weight 212.25
Percent Purity ≥89.0% (GC)
Physical Form Liquid
Chemical Name or Material Neopentyl Glycol Diacrylate (stabilized with MEHQ)
EINECSNumber (2)-1007&(2)-0958
RTECSNumber AS8925000
TSCA Yes