missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl bromide, 98%

Catalog No. AC425720050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425720050 5 g Glass bottle
AC425720250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425720050 Supplier Thermo Scientific Chemicals Supplier No. 425720050

CAS: 630-17-1 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 630-17-1
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000209
InChI Key CQWYAXCOVZKLHY-UHFFFAOYSA-N
Synonym neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g
PubChem CID 12415
IUPAC Name 1-bromo-2,2-dimethylpropane
SMILES CC(C)(C)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.1990g/mL
Boiling Point 105°C to 106°C (767.0 mmHg)
Flash Point 7°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2Br
Quantity 5 g
Specific Gravity 1.199
Formula Weight 151.04
Percent Purity 98%
Chemical Name or Material Neopentyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 211-132-5

RUO – Research Use Only