missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl alcohol, 99%

Catalog No. AAA1439036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1439036 500 g
AAA1439014 25 g
AAA1439022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1439036 Supplier Thermo Scientific Chemicals Supplier No. A1439036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75-84-3 | C5H12O | 88.15 g/mol

It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the Mitsunobu reaction of methyl salicylate coupling of sterically hindered substrates.

Solubility
Soluble in water at (20°C 40 g/L), alcohol, and ether. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 75-84-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004682
InChI Key KPSSIOMAKSHJJG-UHFFFAOYSA-N
Synonym neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
PubChem CID 6404
IUPAC Name 2,2-dimethylpropan-1-ol
SMILES CC(C)(C)CO

Specifications

Melting Point 52°C to 56°C
Density 0.818
Boiling Point 111°C to 114°C
Flash Point 36°C (96°F)
Quantity 500 g
UN Number UN1325
Beilstein 1730984
Merck Index 14,6457
Solubility Information Soluble in water at (20°C 40g/L),alcohol,and ether. Solubility in methanol is almost transparent.
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material Neopentyl alcohol
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-907-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only