accessibility menu, dialog, popup

UserName

CH3CH3H3CHO

CAS RN 75-84-3

IUPAC Name: 2,2-dimethylpropan-1-ol
Synonyms: neopentyl alcohol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: KPSSIOMAKSHJJG-UHFFFAOYSA-N
SMILES: CC(C)(C)CO
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)