missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System

The Neon NxT Electroporation System with 1-Channel Pipette is a next-generation electroporation platform leveraging the unique and trusted Neon electroporation technology, which increases transfection efficiency and cell viability.

Supplier:  Invitrogen™ NEON1S2YR

 View more versions of this product

Catalog No. NEON1S2YR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The Neon NxT Electroporation System is a next-generation electroporation platform leveraging the unique and trusted Neon electroporation technology, which increases transfection efficiency and cell viability. The system streamlines the transfection of mammalian cells, including primary, stem, and difficult-to-transfect cells, for fast and efficient delivery of nucleic acids.

Features of the Neon NxT Electroporation System include:

  • Proven performance—up to 90% transfection efficiency in many cell types, including primary, stem, and difficult-to-transfect cells, while preserving cell viability with electroporation tip technology
  • Flexible—easily transfect 1 x 104 cells to 1 x 107 cells per reaction
  • Simple—easy to perform three-step process with single buffer kit for all cell types that includes Neon NxT Resuspension Genome Editing Buffer
  • Customizable—optimize electroporation parameters freely or leverage over 400 pre-programmed protocols with ability to save up to 10,000 additional protocols
  • Connectivity—Invitrogen TransfectionLab cloud app on Thermo Fisher Connect paired with Instrument Connect allows for digital experiment design and connectivity to Neon NxT system
  • Improved usability—improved pipette ergonomics with ClipTip interlocking technology, enhanced feedback loop, and intuitive user interface with plate setup

Trusted electroporation tip technology

Unlike standard cuvette-based electroporation chambers, the Neon NxT system uses a proprietary, biologically compatible pipette tip chamber that maximizes the distance between the two electrodes while minimizing their surface area, generating a more uniform electric field. This design allows better maintenance of physiological conditions, resulting in high cell survival compared to conventional electroporation.

Sample preservation

The unique Neon NxT pipette tip eliminates the need for a separate electroporation cuvette or plate, while the compact design of the instrument enables an easy fit within a biosafety cabinet. Valuable cells are not lost through sample transfer and are at reduced risk of contamination.

Plug-n-play out of the box

The electroporation process couldn't be simpler. Choose Quick Start or set up a plate for an entire experiment, assigning protocols to each well. Aspirate cells and plasmid mix into the Neon NxT pipette tip, dock into the pipette station, and press electroporate. Watch as the selected well on the screen receives a green checkmark indicating successful electroporation; then pipette the transfected cells into your culture vessel.

Perform more transfections with limited or precious cells

Your cells may be limited when you are required to perform neuronal preps or when enriching for homogeneous cell populations via FACS or Dynabeads enrichment. In these instances, reducing the number of cells required per transfection can allow you to perform more complex experiments with your precious cells.

Neon NxT service options

Standard warranty

Each Neon NxT system comes with a one-year warranty. If the instrument requires repair within the first year, it can be sent to our repair center and repaired at no cost to you. Purchasing one of the below extended warranty options would provide the indicated services during the initial warranty period.

Extended warranty options

  • Rapid Exchange (REX) (Cat. No. ZGEXSCNEONNXT) If your Neon NxT device requires repair, it can be exchanged for a replacement factory-refurbished device at no additional cost, usually arriving in just two business days.
  • ABRC Support (Cat. No. ZG03SCNEONNXT) If your Neon NxT device requires repair, it can be sent for repair and returned to you within 21 business days.
  • ABRC Support Plus (Cat. No. ZG04SCNEONNXT) If your Neon NxT device requires repair, it can be sent for repair and returned to you within 21 business days. And while your device is being repaired, a loaner instrument will be sent to prevent any downtime.

Qualification Services (Cat. No. IQOQ A57744)

You can add qualification services to complement your extended warranty service plan. Instrument hardware qualifications for the Neon NxT system include installation qualification (IQ) and operational qualification (OQ) to document and verify that the instrument is installed and operating according to manufacturer's specifications.

IQ provides documented evidence and verification that the instrument has been delivered and installed according to manufacturer's specifications.

OQ provides documented verification that the instrument subsystems are operating as designed and verifies that the functionality of an instrument meets the manufacturer's operational specifications.

IQOQ is recommended at installation, after moving the instrument, after software or hardware upgrades, and after critical repairs. Our qualification specialists will partner with you to deliver timely, cost-effective, and trusted qualification services that include reliable, audit-style documentation.

Ask your instrument sales representative about acquiring extended warranty coverage and IQOQ at time of instrument purchase.

For those instruments already in your lab, please contact your local service sales representative or visit Contact Us.

Contract options are not available in all regions.

Specifications

Chemical Identifiers

Flasks, Plates (6-well, 12-well, 24-well, and 96-well)
Neon™ NxT Electroporation System
500 to 2500 V (Electroporation)
Color Touchscreen
1 to 100 ms
270 W
• Neon NxT electroporation device
• 1-Channel Pipette
• 1-Channel Pipette Station
• Power cord
• Quick reference guide
Transfection
100/240 VAC
500 to 2500 V (Electroporation)
1 to 10 Pulses
Each
100/240 VAC, 50/60 Hz, 270 VA
Touchscreen

Specifications

Flasks, Plates (6-well, 12-well, 24-well, and 96-well)
Neon™ NxT Electroporation System
500 to 2500 V (Electroporation)
Color Touchscreen
1 to 100 ms
270 W
• Neon NxT electroporation device
• 1-Channel Pipette
• 1-Channel Pipette Station
• Power cord
• Quick reference guide
2-year Extended Warranty, ABRC Support
Transfection
100/240 VAC
500 to 2500 V (Electroporation)
1 to 10 Pulses
Each
100/240 VAC, 50/60 Hz, 270 VA
Touchscreen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.