missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes
Description
The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. No. MPK5000) with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 100 μL, containing 5 x 105 to 2 x 106 adherent cells or 1 x 106 to 5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be plated into a multi-well culture dish.
Specifications
Specifications
| Content And Storage | Box 1 includes: • Resuspension R Buffer, 2 x 80 mL • Resuspension T Buffer, 2 x 80 mL • Resuspension Genome Editing Buffer, 2 x 80 mL • Electrolytic E100 Buffer, 2 x 100 mL Store at 4°C. Box 2 includes: • Reaction tips, 384 tips • Electroporation 8-channel tubes, 16 tubes Store at room temperature. |
| Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Electroporation Pipette Tip |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| For Use With (Application) | Transfection |
| No. of Reactions | 384 x 2 Reactions |
| Quantity | 384 x 2 Reactions |
| Volume (Metric) Sample | 100 μL |
For Research Use Only. Not for use in diagnostic procedures.