missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.

Supplier:  Invitrogen™ N10968

 View more versions of this product

Catalog No. N10968


Only null left
Explore available promotions

Description

Description

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. No. MPK5000) with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 10 μL, containing 5 x 104 to 2 x 105 adherent cells or 1 x 105 to 5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be plated into a multi-well culture dish.

Specifications

Specifications

Stem Cell, Primary Cell, Hard-to-Transfect Cell
Electroporation Pipette Tip
Neon™ NxT Electroporation System
96 x 2 Reactions
10 μL
Box 1 includes:
• Resuspension R Buffer, 4 x 1 mL
• Resuspension T Buffer, 4 x 1 mL
• Resuspension Genome Editing Buffer, 4 x 1 mL
• Electrolytic E10 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 96 tips
• Electroporation 8-channel tubes, 4 tubes

Store at room temperature.
Electroporation
Transfection
96 x 2 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.