missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neon™ NxT Electroporation System 10-μL Kit with 1-Channel Tubes
Description
The Neon NxT Electroporation System 10-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 10 μL, containing 5 x 104–2 x 105 adherent cells or 1 x 105–5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be washed and plated into a multi-well culture dish.
Specifications
Specifications
| Content And Storage | • 1 mL Resuspension R Buffer; store at 4°C • 1 mL Resuspension T Buffer; store at 4°C • 1 mL Resuspension Genome Editing Buffer; store at 4°C • 50 mL Electrolytic E10 Buffer; store at 4°C • 25 reaction tips • 8 electroporation 1-channel tubes |
| Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Electroporation Pipette Tip |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| For Use With (Application) | Transfection |
| No. of Reactions | 25 x 2 Reactions |
| Product Type | Pipette Tip |
| Quantity | 25 x 2 reactions |
| Volume (Metric) Sample | 10 μL |
For Research Use Only. Not for use in diagnostic procedures.