missing translation for 'onlineSavingsMsg'
Learn More

Neodymium Standard For ICP/MS, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLND21BY

Catalog No. CLND21BY

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Nd1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Neodymium Standard
7440-00-8
Nd
ICP-MS
Standard Solution
QEFYFXOXNSNQGX-UHFFFAOYSA-N
2% HNO3
144.24
23934
1mg/L
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011130
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ICP/ICP-MS Standard
[Nd]
neodymium
125 mL
CHEBI:33372

Chemical Identifiers

7440-00-8
144.24
QEFYFXOXNSNQGX-UHFFFAOYSA-N
CHEBI:33372
[Nd]
Nd
MFCD00011130
23934
neodymium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only