missing translation for 'onlineSavingsMsg'
Learn More

Neodymium, plasma standard solution, Specpure™ Nd 1000μg/mL

$100.45 - $100.45

Chemical Identifiers

MDL Number MFCD00011130
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13882AD
View Documents
Thermo Scientific Chemicals
013882AD
50 mL
Each for $100.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00011130

Specifications

Liquid
MFCD00011130
50 mL
Neodymium plasma standard solution, Nd 1000μg/mL
Nd2O3 in 5% HNO3
UN3264
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only