missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neodymium(III) Trifluoromethanesulfonate 97.0+%, TCI America™
$91.33 - $290.79
Chemical Identifiers
| CAS | 34622-08-7 |
|---|---|
| Molecular Formula | C3F9NdO9S3 |
| Molecular Weight (g/mol) | 591.432 |
| MDL Number | MFCD00192343 |
| InChI Key | WYRSPTDNOIZOGA-UHFFFAOYSA-K |
| Synonym | neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate |
| PubChem CID | 2733937 |
| IUPAC Name | neodymium(3+);trifluoromethanesulfonate |
| SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] |
Chemical Identifiers
| 34622-08-7 | |
| 591.432 | |
| WYRSPTDNOIZOGA-UHFFFAOYSA-K | |
| 2733937 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] |
| C3F9NdO9S3 | |
| MFCD00192343 | |
| neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate | |
| neodymium(3+);trifluoromethanesulfonate |
Specifications
| 34622-08-7 | |
| C3F9NdO9S3 | |
| 5 g | |
| neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3] | |
| 591.432 | |
| 591.43 | |
| Crystalline Powder |
| Purple | |
| MFCD00192343 | |
| 1759 | |
| WYRSPTDNOIZOGA-UHFFFAOYSA-K | |
| neodymium(3+);trifluoromethanesulfonate | |
| 2733937 | |
| ≥97.0% (T) | |
| Neodymium(III) Trifluoromethanesulfonate |
Safety and Handling
TSCA : No