missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

$161.16 - $161.16

Chemical Identifiers

CAS 34622-08-7
Molecular Formula C3F9NdO9S3
Molecular Weight (g/mol) 591.432
MDL Number MFCD00192343
InChI Key WYRSPTDNOIZOGA-UHFFFAOYSA-K
Synonym neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
PubChem CID 2733937
IUPAC Name neodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4339309
View Documents
Thermo Scientific Chemicals
04339309
10 g
Each for $161.16
Only null left
 
Description

Description

Neodymium(III) trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes and as a catalyst for dehydration of fructose, Diels-Alder reactions of cyclopentadiene and alkyl acrylates, α-amination reactions of pyrazolones, polymerization reactions of cyclosiloxanes, direct polycondensation of dicarboxylic acids and diols, regioselective reductive ring opening of benzylidene acetals, Paal-Knorr cyclocondensation and nitration of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34622-08-7
591.432
WYRSPTDNOIZOGA-UHFFFAOYSA-K
2733937
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]
C3F9NdO9S3
MFCD00192343
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
neodymium(3+);trifluoromethanesulfonate

Specifications

34622-08-7
10 g
MFCD00192343
Soluble in water.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]
591.432
591.44
1.7 g/mL
Powder
C3F9NdO9S3
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
WYRSPTDNOIZOGA-UHFFFAOYSA-K
neodymium(3+);trifluoromethanesulfonate
2733937
98%
Neodymium (III) trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only