missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Catalog No. AA40568NB
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40568NB 5 g
AA40568ND 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40568NB Supplier Thermo Scientific Chemicals Supplier No. 040568NB

CAS: 17522-69-9 | Cl3NdO12 | 442.58 g/mol

Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17522-69-9
Molecular Formula Cl3NdO12
Molecular Weight (g/mol) 442.58
MDL Number MFCD00198195
InChI Key HBKMAYJLXKBOER-UHFFFAOYSA-K
Synonym neodymium 3+ perchlorate, neodymium perchlorate, neodymium 3+ triperchlorate, neodymium 3+ ion triperchlorate ion, neodymium iii perchlorate aqueous solution
PubChem CID 13783607
IUPAC Name neodymium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Nd+3]

Specifications

Physical Form Liquid
Quantity 5 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 550.68 (442.59 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Neodymium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 236-821-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only