accessibility menu, dialog, popup

UserName

OClOOOOClOOOOClOOONd

CAS RN 17522-69-9

IUPAC Name: neodymium(3+) triperchlorate
Synonyms: neodymium(3+) triperchlorate
Molecular Weight (g/mol): 442.58
Molecular Formula: Cl3NdO12
InChi Key: HBKMAYJLXKBOER-UHFFFAOYSA-K
SMILES: [Nd+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results