missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine Hydrochloride Monohydrate, 99%

CAS: 303136-82-5 | C14H12N2 | 208.26 g/mol

Supplier:  Thermo Scientific Chemicals 153310250

 View more versions of this product

Catalog No. AC153310250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Neocuproine hydrochloride monohydrate
98.5
250°C
Authentic
Glass Bottle
MFCD00150062
2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 %
CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
208.26
262.75
Fine Crystalline Powder
303136-82-5,484-11-7
100.0
Yellow
99%
C14H12N2
25 g
IYRGXJIJGHOCFS-UHFFFAOYSA-N
2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride
2723838
99%

Chemical Identifiers

303136-82-5
208.26
IYRGXJIJGHOCFS-UHFFFAOYSA-N
2723838
CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
C14H12N2
MFCD00150062
2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 %
2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only