missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine Hydrochloride Monohydrate, 99%

Catalog No. p-3737051
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153310050 5 g
AC153310250 25 g
AC153311000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC153310050 Supplier Thermo Scientific Chemicals Supplier No. 153310050

CAS: 303136-82-5 | C14H12N2 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 303136-82-5 , 484-11-7
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00150062
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym 2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 %
PubChem CID 2723838
IUPAC Name 2,9-dimethyl-1,10-phenanthroline;hydrate;hydrochloride
SMILES CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 250°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 262.75
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Neocuproine hydrochloride monohydrate
Show More Show Less

RUO – Research Use Only