missing translation for 'onlineSavingsMsg'
Learn More

Neoandrographolide, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013791
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013791 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013791 Supplier Merck Emd Millipore Supplier No. 498795MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 27215-14-1
Physical Form Neat
Linear Formula C26H40O8
MDL Number MFCD07779130
Synonym ent-19-Hydroxy-8(17),13-labdadien-16,15-olide 19-O-beta-D-glucopyranoside; D-Glucopyranosyloxy)methyl]decahydro-5,8 A-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5 H)-furanone
Type Analytical Standard
Quantity 5 mg
Formula Weight 480.59
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less