missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nelfinavir Mesylate Hydrate 97.0+%, TCI America™
$386.54 - $688.49
Chemical Identifiers
CAS | 159989-65-8 |
---|---|
Molecular Formula | C33H49N3O7S2 |
Molecular Weight (g/mol) | 663.889 |
MDL Number | MFCD00931436 |
InChI Key | NQHXCOAXSHGTIA-SKXNDZRYSA-N |
Synonym | nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777 |
PubChem CID | 64142 |
ChEBI | CHEBI:7497 |
IUPAC Name | (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid |
SMILES | CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
N098625MG
|
TCI America
N098625MG |
25 mg |
Each for $386.54
|
|
|||||
N0986100MG
|
TCI America
N0986100MG |
100 mg |
Each for $688.49
|
|
|||||
Chemical Identifiers
159989-65-8 | |
663.889 | |
NQHXCOAXSHGTIA-SKXNDZRYSA-N | |
64142 | |
(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid |
C33H49N3O7S2 | |
MFCD00931436 | |
nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777 | |
CHEBI:7497 | |
CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O |
Specifications
159989-65-8 | |
C33H49N3O7S2 | |
25 mg | |
NQHXCOAXSHGTIA-SKXNDZRYSA-N | |
(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid | |
64142 | |
663.89 | |
Crystalline Powder |
White-Yellow | |
MFCD00931436 | |
nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777 | |
CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O | |
663.889 | |
CHEBI:7497 | |
≥97.0% (HPLC) | |
Nelfinavir Mesylate Hydrate |
Safety and Handling
RTECSNumber : NW6995333
TSCA : No
Recommended Storage : Refrigerator