Learn More
Invitrogen™ Human NEK7, His Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PR7441A
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic Domain
NEK7 (NIMA Related Kinase 7) is a Protein Coding gene. Diseases associated with NEK7 include Muckle-Wells Syndrome and Cinca Syndrome. Among its related pathways are Cell Cycle, Mitotic and Transport of the SLBP independent Mature mRNA. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is NEK6.Specifications
| Q8TDX7 | |
| Kinase Assay | |
| 140609 | |
| Human NEK7, His Tag | |
| RUO - research use only | |
| NEK7 | |
| 8-313 | |
| Human | |
| His-tag | |
| Insect cells |
| See Label | |
| 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 | |
| 63.7 kDa | |
| 100 μg | |
| 2810460C19Rik; AU020186; NEK7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; nimA-related protein kinase 7; Serine/threonine-protein kinase Nek7 | |
| NEK Kinase Family | |
| Unconjugated | |
| Recombinant | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Liquid |