missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Needles Array Electrode Handles for BTX™ AgilePulse™ in Vivo

Needle Array Handle for miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ in vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.

$3576.10 - $3576.10

Specifications

For Use With (Application) Gene and Vaccine Delivery
Material Plastic
Product Line BTX Electroporation
Product Type Needle Array Handle
Includes Handle Only
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
BTX470000
View Documents
BTX™
470000
Generators with SN AP-4068-ID/AP-5030-IM and Before
Each for $3,576.10
Only null left
 
Description

Description

The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.

The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.

  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from
Specifications

Specifications

Gene and Vaccine Delivery
BTX Electroporation
Handle Only
Non-sterile
Square Wave
Plastic
Needle Array Handle
1 Ea.
Electrode Connector Cable
Videos
SDS
Documents

Documents