missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEDL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101569
Description
Antibody detects endogenous levels of total HECW2.
E3 ubiquitin-protein ligase that mediates ubiquitination of TP73. Acts to stabilize TP73 and enhance activation of transcription by TP73.Chemical Identifiers
| NEDL2 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| HECW2 | |
| A730039N16Rik; D030049F17Rik; E3 ubiquitin-protein ligase HECW2; Gm971; hect; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2, isoform 2; HECT, C2 and WW domain-containing protein 2; HECT-type E3 ubiquitin transferase HECW2; HECW2; KIAA1301; mKIAA1301; NEDD4-like E3 ubiquitin-protein ligase 2; NEDD4-related E3 ubiquitin ligase NEDL2; NEDL2 | |
| HECW2 | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P2P5 | |
| 57520 | |
| A synthesized peptide derived from human HECW2(Accession Q9P2P5), corresponding to amino acid residues E485-E535. | |
| Antibody |
Specifications
| NEDL2 | |
| Polyclonal | |
| Unconjugated | |
| HECW2 | |
| A730039N16Rik; D030049F17Rik; E3 ubiquitin-protein ligase HECW2; Gm971; hect; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2; HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2, isoform 2; HECT, C2 and WW domain-containing protein 2; HECT-type E3 ubiquitin transferase HECW2; HECW2; KIAA1301; mKIAA1301; NEDD4-like E3 ubiquitin-protein ligase 2; NEDD4-related E3 ubiquitin ligase NEDL2; NEDL2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57520 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P2P5 | |
| HECW2 | |
| A synthesized peptide derived from human HECW2(Accession Q9P2P5), corresponding to amino acid residues E485-E535. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |