missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEDD4 Monoclonal Antibody (K24022_16E6), Invitrogen™

Catalog No. MA555813
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555813 50 μL
MA556193 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555813 Supplier Invitrogen™ Supplier No. MA555813
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: NARLTIFGNS AVSQPASSSN HSSRRGSLQA YTFEEQPTLP VLLPTSSGLP PGWEEKQDER GRSYYVDHNS RTTTWTKPTV QATVETSQLT SSQSSAGPQS QASTSDSGQQ

The E3 ubiquitin-protein ligase NEDD4 is a Ubl conjugation pathway protein. It contains one HECT (E6AP-type E3 ubiquitin-protein ligase) domain and four WW domains. It is involved in the pathway leading to the degradation of VEGFR-2/KDFR, independently of its ubiquitin-ligase activity. It acts as a potential proto-oncogene that negatively regulates PTEN via ubiquitination, a paradigm analogous to that of Mdm2 and p53. It is also reported to be involved in the budding of many viruses and is the regulator of epithelial Na+ channels, which lead to arterial hypertension. NEDD4 protein is localized to the sarcolemmal region of muscle fibers. Increase in NEDD4 protein expression leads to unloaded and denervated muscles as well as a decrease in Notch1 expression. Hence it acts as down regulator of Notch-1 gene expression.

Specifications

Antigen NEDD4
Applications Western Blot
Classification Monoclonal
Clone K24022_16E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NEDD4
Gene Accession No. P46934
Gene Alias AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1
Gene Symbols NEDD4
Host Species Mouse
Immunogen Recombinant human NEDD4 (immunogen range is 721-830).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less