missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115632 Supplier Invitrogen™ Supplier No. PA5115632
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NDUFS6.

This gene encodes a subunit of the NADH:ubiquinone oxidoreductase, which is the first enzyme complex in the electron transport chain of mitochondria. This complex functions in the transfer of electrons from NADH to the respiratory chain. The subunit encoded by this gene is one of seven subunits in the iron-sulfur protein fraction. Mutations in this gene cause mitochondrial complex I deficiency, a disease that causes a wide variety of clinical disorders, including neonatal disease and adult-onset neurodegenerative disorders.

Specifications

Antigen NDUFS6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NDUFS6
Gene Accession No. O75380, P52503, P52504
Gene Alias BC059730; CI-13kA; CI13KDA; CI-13kD-A; complex I 13kDa subunit A; complex I, mitochondrial respiratory chain, 13-kD subunit; complex I-13kD-A; IP13; Ip13dis; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) Fe-S protein 6; NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial; NADH dehydrogenase Fe-S protein 6; NADH ubiquinone oxidoreductase; NADH:ubiquinone oxidoreductase NDUFS6 subunit; NADH:ubiquinone oxidoreductase subunit S6; NADH-ubiquinone oxidoreductase 13 kDa-A subunit; Ndub13; NDUFS6; RATIp13dis; zgc:103774
Gene Symbols NDUFS6
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDUFS6(Accession O75380), corresponding to amino acid residues S33-R83.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29478, 407785, 4726
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less