missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFB8 Monoclonal Antibody (K9N014_11C6), Invitrogen™

Catalog No. MA556007
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556007 200 μL
MA555627 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556007 Supplier Invitrogen™ Supplier No. MA556007
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: YPRTPEERAA AAKKYNMRVE DYEPYPDDGM GYGDYPKLPD RSQHERDPWY SWDQPGLRLN WGEPMHWHLD

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [UniProt].

Specifications

Antigen NDUFB8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K9N014_11C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NDUFB8
Gene Accession No. O95169, Q9D6J5
Gene Alias 19 kDa; 2900010I05Rik; AI987932; ASHI; CI-ASHI; complex I ASHI subunit; complex I-ASHI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase 1 beta subcomplex 8; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase subunit B8; NADH-ubiquinone oxidoreductase ASHI subunit; NDUFB8
Gene Symbols NDUFB8
Host Species Mouse
Immunogen Recombinant human NDUFB8 (immunogen range is 41-110).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293991, 4714, 67264
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less