missing translation for 'onlineSavingsMsg'
Learn More

NDUFB11 Antibody, ProSci

Catalog No. 16813440
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16813440 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16813440 Supplier ProSci Llc Supplier No. 161380100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

NDUFB11 Polyclonal Antibody specifically detects RPS27L in Human, Mouse, Rat Samples. It is validated for Western Blot.

Specifications

Antigen NDUFB11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution WB: 1:500 - 1:2000
Formulation PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
Gene Alias CI-ESSS, ESSS, MC1DN3NP17.3, Np15, P17.3, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa, NADH-ubiquinone oxidoreductase ESSS subunit, complex I NP17.3 subunit, complex I-ESSS, neuronal protein 17.3
Host Species Rabbit
Purification Method Affinity purification
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54539
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze / thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Neuroscience
Show More Show Less