missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFA8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA559604

Catalog No. PIPA559604

Only null left

Description

Description

Immunogen sequence: YWTCIDYTGQ QLFRHCRKQQ AKFDECVLDK LGWVRPDLGE LSKVTKVKTD RPLPENPYHS RPRPDPSPEI EGDLQPATHG SRFYFWTK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 88%.

NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Chemical Identifiers

NDUFA8
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse, Rat
NDUFA8
0610033L03Rik; Aa2-258; AW261656; CI-19KD; CI-PGIV; Complex I-19kD; complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH:ubiquinone oxidoreductase PGIV subunit; NADH:ubiquinone oxidoreductase subunit A8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NDUFA8; PGIV
NDUFA8
Primary
Antigen affinity chromatography
0.05 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P51970, Q9DCJ5
296658, 4702, 68375
Recombinant protein corresponding to Human NDUFA8. Recombinant protein control fragment (Product #RP-98407).
Antibody

Specifications

NDUFA8
Polyclonal
Unconjugated
NDUFA8
0610033L03Rik; Aa2-258; AW261656; CI-19KD; CI-PGIV; Complex I-19kD; complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH:ubiquinone oxidoreductase PGIV subunit; NADH:ubiquinone oxidoreductase subunit A8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NDUFA8; PGIV
Rabbit
Antigen affinity chromatography
RUO
296658, 4702, 68375
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P51970, Q9DCJ5
NDUFA8
Recombinant protein corresponding to Human NDUFA8. Recombinant protein control fragment (Product #RP-98407).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions