missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ NDSB-201, Calbiochem™,
Supplier: MilliporeSigma™ 480005250GM
| Quantity | 250 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 15471-17-7 | |
| 201.24 | |
| REEBJQTUIJTGAL-UHFFFAOYSA-N | |
| 84929 | |
| [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 |
| C8H11NO3S | |
| MFCD00064468 | |
| 3-(1-Pyridino)-1-propane Sulfonate | |
| 1-(3-sulfonatopropyl)pyridin-1-ium |
Specifications
| White | |
| 250 g | |
| C8H11NO3S | |
| 3-(1-Pyridino)-1-propane Sulfonate | |
| [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 | |
| 201.24 | |
| ≥99% (Titration) |
| 15471-17-7 | |
| Glass Bottle | |
| MFCD00064468 | |
| REEBJQTUIJTGAL-UHFFFAOYSA-N | |
| 1-(3-sulfonatopropyl)pyridin-1-ium | |
| 84929 |