missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NDSB-201, Calbiochem™,

Catalog No. p-7082966
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
48000525GM 25 g Glass Bottle
480005250GM 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48000525GM Supplier MilliporeSigma™ Supplier No. 48000525GM
Only null left

Chemical Identifiers

CAS 15471-17-7
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00064468
InChI Key REEBJQTUIJTGAL-UHFFFAOYSA-N
Synonym 3-(1-Pyridino)-1-propane Sulfonate
PubChem CID 84929
IUPAC Name 1-(3-sulfonatopropyl)pyridin-1-ium
SMILES [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Packaging Glass Bottle
Percent Purity ≥99% (Titration)