missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCKAP1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558813
Description
Immunogen sequence: VLIRMYNIKK TCSDPKSKPP FLLEKSMEPS LKYINKKFPN IDVRNSTQHL GPVHREKAEI IRFLTNYYQS FVD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
This gene encodes a member of the HEM family of tissue-specific transmembrane proteins which are highly conserved from invertebrates through mammals. This gene is only expressed in hematopoietic cells. The encoded protein is a part of the Scar/WAVE complex which plays an important role in regulating cell shape in both metazoans and plants. Alternatively spliced transcript variants encoding different isoforms have been found.[provided by RefSeq, May 2010].Chemical Identifiers
| NCKAP1L | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| NCKAP1L | |
| 4930568P13Rik; AI463083; HEM1; Hematopoietic protein 1; Hemp1; membrane-associated protein hem-1; NCK associated protein 1 like; NCKAP1L; nck-associated protein 1-like | |
| NCKAP1L | |
| Primary | |
| Antigen affinity chromatography |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55160 | |
| 3071 | |
| Recombinant protein corresponding to Human NCKAP1L. Recombinant protein control fragment (Product #RP-97719). | |
| Antibody |
Specifications
| NCKAP1L | |
| Polyclonal | |
| Unconjugated | |
| NCKAP1L | |
| 4930568P13Rik; AI463083; HEM1; Hematopoietic protein 1; Hemp1; membrane-associated protein hem-1; NCK associated protein 1 like; NCKAP1L; nck-associated protein 1-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55160 | |
| NCKAP1L | |
| Recombinant protein corresponding to Human NCKAP1L. Recombinant protein control fragment (Product #RP-97719). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |