missing translation for 'onlineSavingsMsg'
Learn More

NBD-Cl (=4-Chloro-7-nitro-2,1,3-benzoxadiazole) 98.0+%, TCI America™

Catalog No. p-7118288
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55921G 1 g
A55925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55921G Supplier TCI America Supplier No. A55921G
Only null left

Chemical Identifiers

CAS 10199-89-0
Molecular Formula C6H2ClN3O3
Molecular Weight (g/mol) 199.55
MDL Number MFCD00005808
InChI Key IGHBXJSNZCFXNK-UHFFFAOYSA-N
Synonym 4-chloro-7-nitrobenzofurazan, nbd chloride, 4-chloro-7-nitrobenzo-2-oxa-1,3-diazole, nbd-cl, 7-chloro-4-nitrobenzofurazan, 2,1,3-benzoxadiazole, 4-chloro-7-nitro, nbd-chloride, nbd-c 1, 1-chloro-4-nitrobenzoxadiazole, 4-nitro-7-chlorobenzofurazan
PubChem CID 25043
ChEBI CHEBI:78878
IUPAC Name 4-chloro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 98°C
Color Yellow
Quantity 1 g
Formula Weight 199.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material NBD-Cl (=4-Chloro-7-nitro-2,1,3-benzoxadiazole) [for HPLC Labeling]
RTECSNumber DF8002400
TSCA Yes