accessibility menu, dialog, popup

UserName

ONOClNON

CAS RN 10199-89-0

IUPAC Name: 4-chloro-7-nitro-2,1,3-benzoxadiazole
Synonyms: 4-chloro-7-nitro-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 199.55
Molecular Formula: C6H2ClN3O3
InChi Key: IGHBXJSNZCFXNK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C2=NON=C12
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)