missing translation for 'onlineSavingsMsg'
Learn More

NBD chloride, 98%

Catalog No. p-171603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172390010 1 g Glass bottle
AC172390050 5 g Glass bottle
AC172390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC172390010 Supplier Thermo Scientific Chemicals Supplier No. 172390010

missing translation for 'validMainframeMsg'

CAS: 10199-89-0 | C6H2ClN3O3 | 199.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10199-89-0
Molecular Formula C6H2ClN3O3
Molecular Weight (g/mol) 199.55
MDL Number MFCD00005808
InChI Key IGHBXJSNZCFXNK-UHFFFAOYSA-N
Synonym 4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitrobenzo-2-oxa-1, 3-diazole
PubChem CID 25043
ChEBI CHEBI:78878
IUPAC Name 4-chloro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 96.0°C to 99.0°C
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Color Brown to Yellow
Solubility Information Solubility in water: soluble.
Quantity 1 g
Physical Form Crystalline Powder
Formula Weight 199.55
Percent Purity 98%
Chemical Name or Material NBD chloride
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-496-4
RTECSNumber DF8002400
TSCA TSCA

RUO – Research Use Only