Learn More
OHAUS™ Navigator Portable Precision Balance
The Navigator series offers essential weighing applications at a competitive price point.
Supplier:  OHAUS™ 30642239
 View more versions of this product
Catalog No. 01920285
Only null left
Description
Description
  • The Navigator series has advanced weighing technology which offers one-second stabilization for fast and accurate weighing results. This improves operator efficiency, productivity and throughput.
  • Navigators have overload protection systems that can withstand loads up to four times the rated capacity safeguards the balance.
Specifications
Specifications
IEC/EN 61010-1, IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B; Canada ICES-003 Class B, CE, RCM
220 g
0.001 g
ABS, Stainless Steel
2.5 sec.
230 x 204 x 104 mm
AC adapter (included) or 4 C batteries (not included)
RS232, USB, or Ethernet (available as accessories)
LCD
0.005 g
NV223
Toploading Balances
1 kg
Product Suggestions
Product Suggestions