missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Natural LDPE Boston Round Bottles with Black Phenolic Caps

Natural LDPE Boston Round Bottles are very flexible with high impact strength. Excellent for mild and strong buffers, good chemical resistance

 View more versions of this product

Catalog No. FB02911966A


Only null left

Description

Description

  • Bottles retain their impact strength at very low temperatures
  • Caps form an tight seal and offer a good chemical barrier
  • Liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
Specifications

Chemical Identifiers

Natural
Black
Narrow
Polyethylene Cone
LDPE
Yes
Boston Round Bottle
Phenolic

Specifications

120 mL
Natural
17 mm
LDPE
Black
Yes
106 mm
Polyethylene Cone
24-410 mm
4 oz.
22 mm
109 mm
48
24-410
Narrow
Boston Round Bottle
Phenolic
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions