missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Natural HDPE Heavy Weight Jug with F422 Foam-lined PP Cap

Catalog No. 01677104
Change view
Click to view available options
Capacity (Metric):
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01677104 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01677104 Supplier Qorpak™ Supplier No. 329124

Heavy-weight, 190 gram jug has an integrated handle and 38-439 neck finish for easy and drip-free pouring. 4 jugs per DOT-UN Approved Reshipper Carton.

  • Used in a wide variety of applications including: electronic chemical, laboratory chemical, specialty chemical and general chemical industries
  • Designed for high purity applications where particle contamination requirements are stringent
  • The U.N. Certified 4G/Y33.7/S package is used for packaging acid, alkaline and solvent hazardous chemicals having a specific gravity of 1.9 or less
  • Product is packed with an F422 PE foam-lined polypropylene cap to maintain cleanliness inside bottle

Specifications

Capacity (Metric) 4 L
Color Natural
Diameter (English) 6 in.
Material HDPE
Packaging Carton
Quantity 4/Pk.
Sterility Non-sterile
Weight (Metric) 190 g
Width (Metric) 15.24 cm
Closure Color White
Closure Included Yes
Graduated No
Mouth Narrow, Threaded
Height (Metric) 34.5 cm
Width (English) 6 in.
Height (English) 13.6 in.
Closure Liner F422 Polyethylene Foam
Closure Material Polypropylene
Closure Size 38-439
Diameter (Metric) 15.24 cm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Capacity (English) 135 oz.
Shape Round Jug
Certifications/Compliance DOT-UN Approved Reshipper Carton
Disposable Reusable
Product Type Jug and Lid
Show More Show Less