Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Catalog No. 0339691
Change view
Click to view available options
Marking Spot:
No
Closure Color:
Light Blue
Septum:
PTFE/Silicone, Pre-slit
Color:
Clear
Catalog No. Marking Spot Closure Color Septum Color
0339691 No Light Blue PTFE/Silicone, Pre-slit Clear
1 options
Catalog No. 0339691 Supplier Thermo Scientific™ Supplier No. C4010018
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order
Specifications
Closure Color Light Blue
Description Clear Glass; Light Blue Cap; PTFE/Silicone Pre-slit septum
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Pre-slit Yes
Septum PTFE/Silicone, Pre-slit
Marking Spot No