Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Supplier:  Thermo Scientific™ C4013100SS
 View more versions of this product
Catalog No. 03378611
Get Consumables with Purchase
View all promotions
Only null left
Description
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
Specifications
Chemical Identifiers
1 Ea.
Industrial Chromatography
National
Specifications
1 Ea.
Aluminum Seal
Industrial Chromatography
Manual Crimper
13 mm
National
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.