missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits

Catalog No. 03376460
Change view
Click to view available options
Color:
Amber
Closure Type:
Polypropylene, Flanged
Septum:
PTFE/Silicone
Packaging Type:
Storage Tray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Septum Packaging Type
03376460 Amber Polypropylene, Flanged PTFE/Silicone Storage Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03376460 Supplier Thermo Scientific™ Supplier No. C401358AW

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted

Includes: 

100 vials with pre-attached caps and septa

Specifications

Closure Color Black
Color (Seal) Red/White
Description Amber Glass; ID patch; PP Black Cap; Red PTFE/White Silicone septum; in storage tray
Color Amber
Material Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum PTFE/Silicone
Packaging Type Storage Tray
Closure Material Polypropylene
Closure Type Polypropylene, Flanged
Closure Size 8-425
Show More Show Less